2-Methylbenzothiazole 98%

Supplier: Ambeed
Warning

A228257-100G A228257-10G A228257-25G A228257-5G
77275-688EA 70 USD
77275-688 77275-690 77275-692 77275-694
2-Methylbenzothiazole 98%
2-Methylbenzothiazole

Precaución: for Research Use Only

Formula: C₈H₇NS
MDL Number: MFCD00005794
CAS Number: 120-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR