5-(Methoxycarbonyl)-2-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A228150-1G A228150-250MG
77275-678EA 100 USD
77275-678 77275-680
5-(Methoxycarbonyl)-2-nitrobenzoic acid 95%
5-(Methoxycarbonyl)-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₆
MDL Number: MFCD08690063
CAS Number: 76143-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR