2-Chloro-5-nitro-N-(4-pyridyl)benzamide 98%

Supplier: Ambeed
Warning

Synonyms: T0070907, 2-Chloro-5-nitro-N-(pyridin-4-yl)benzamide

A227971-100MG A227971-10MG A227971-1G A227971-1MG A227971-25MG A227971-50MG A227971-5MG
77261-488EA 43 USD
77261-488 77261-490 77261-492 77261-494 77261-496 77261-498 77261-500
2-Chloro-5-nitro-N-(4-pyridyl)benzamide 98%
2-Chloro-5-nitro-N-(4-pyridyl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₈ClN₃O₃
MDL Number: MFCD00121849
CAS Number: 313516-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR