2-(Boc-amino)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Boc-amino)phenylboronic Acid Pinacol Ester

A227968-1G A227968-250MG A227968-25G
77288-306EA 14 USD
77288-306 77288-308 77288-310
2-(Boc-amino)benzeneboronic acid pinacol ester 98%
2-(Boc-amino)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₄
MDL Number: MFCD03411943
CAS Number: 159624-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR