Methanesulfonato(tri-t-butylphosphino)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%
Supplier: Ambeed
|
Warning
|
A227945-100MG
A227945-1G
A227945-250MG
A227945-5G
77249-504EA
47.38
USD
77249-504
77249-506
77550-472
77550-474
Methanesulfonato(tri-t-butylphosphino)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(tri-t-butylphosphino)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II)
Precaución: for Research Use Only
Formula:
C₂₆H₄₂NO₃PPdS MW: 586.07 g/mol |
MDL Number:
MFCD28411657 CAS Number: 1621274-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...