3-(4-Piperidinyl)-3,4-dihydro-2(1H)-quinazolinone 95%

Supplier: Ambeed
Warning

Synonyms: 3-Piperidin-4-yl-3,4-dihydro-1H-quinazolin-2-one, 3-(Piperidin-4-yl)-1,2,3,4-tetrahydroquinazolin-2-one

A227883-1G A227883-250MG
77275-666EA 361 USD
77275-666 77275-668
3-(4-Piperidinyl)-3,4-dihydro-2(1H)-quinazolinone 95%
3-(4-Piperidinyl)-3,4-dihydro-2(1H)-quinazolinone

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₃O
MDL Number: MFCD11043062
CAS Number: 79098-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR