2-Bromo-5-methylpyrazine 98%

Supplier: Ambeed
Warning

A227758-250MG A227758-1G
77261-486EA 59.74 USD
77261-486 77615-896
2-Bromo-5-methylpyrazine 98%
2-Bromo-5-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD08705754
CAS Number: 98006-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR