Isoquinoline-5-boronic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Isoquinolin-5-ylboronic acid hydrochloride

A227451-100MG A227451-1G A227451-250MG
77261-472EA 39 USD
77261-472 77261-474 77261-476
Isoquinoline-5-boronic acid hydrochloride 95%
Isoquinoline-5-boronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₈BNO₂·ClH
MDL Number: MFCD08235064
CAS Number: 1256345-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR