3-(3,4-Dimethoxyphenyl)propan-1-ol 97%

Supplier: Ambeed
Warning

A227368-100MG A227368-1G
77275-656EA 20.6 USD
77275-656 77275-658
3-(3,4-Dimethoxyphenyl)propan-1-ol 97%
3-(3,4-Dimethoxyphenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
MW: 196.24 g/mol
MDL Number: MFCD00002951
CAS Number: 3929-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR