2,4-Dichloro-3-nitroquinoline 95%

Supplier: Ambeed
Warning

A227279-100MG A227279-1G A227279-250MG A227279-5G
77249-490EA 21 USD
77249-490 77249-492 77249-494 77746-334
2,4-Dichloro-3-nitroquinoline 95%
2,4-Dichloro-3-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₄Cl₂N₂O₂
MDL Number: MFCD05666285
CAS Number: 132521-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR