Methyl 2-bromo-4-hydroxybenzoate 97%

Supplier: Ambeed
Warning

A227277-100MG A227277-250MG A227277-1G
77261-468EA 89.61 USD
77261-468 77261-470 77568-202
Methyl 2-bromo-4-hydroxybenzoate 97%
Methyl 2-bromo-4-hydroxybenzoate

Precaución: For research use only.

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
MDL Number: MFCD18398783
CAS Number: 101085-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR