2',3',5'-Tri-O-acetyluridine 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate, Uridine triacetate, Uridine 2',3',5'-triacetate

A227210-100G A227210-5G A227210-25G
77275-644EA 77 USD
77275-644 77275-646 77748-377 77748-376
2',3',5'-Tri-O-acetyluridine 97%
2',3',5'-Tri-O-acetyluridine

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₉
MDL Number: MFCD00023795
CAS Number: 4105-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR