tert-Butyl hydroxy(methyl)carbamate 97%

Supplier: Ambeed
Warning

A227046-10G A227046-250MG A227046-25G A227046-5G A227046-1G
77261-458EA 104 USD
77261-458 77261-460 77261-462 77261-464 77568-196
tert-Butyl hydroxy(methyl)carbamate 97%
tert-Butyl hydroxy(methyl)carbamate

Precaución: For research use only.

Formula: C₆H₁₃NO₃
MW: 147.17 g/mol
MDL Number: MFCD24465554
CAS Number: 19689-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR