2-Anilinopyridine 97%
Supplier: Ambeed
|
Warning
|
A226990-100G
A226990-10G
A226990-1G
A226990-250MG
A226990-5G
77261-448EA
347
USD
77261-448
77261-450
77261-452
77261-454
77261-456
2-Anilinopyridine 97%
2-Anilinopyridine
Precaución: for Research Use Only
Formula:
C₁₁H₁₀N₂ |
MDL Number:
MFCD00006248 CAS Number: 6631-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...