4-Amino-2-(trifluoromethyl)benzamide 97%

Supplier: Ambeed
Warning

A226804-1G A226804-250MG
77261-442EA 240 USD
77261-442 77261-444
4-Amino-2-(trifluoromethyl)benzamide 97%
4-Amino-2-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O
MDL Number: MFCD27933529
CAS Number: 934600-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR