N-(tert-Butoxycarbonyl)-4-aminobutyric acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-GABA-OH, N-Boc-γ-aminobutyric acid, 4-(Boc-amino)butyric Acid, 4-(tert-Butoxycarbonylamino)butyric Acid, N-Boc-4-aminobutyric Acid, N-BOC-gamma-Aminobutyric acid

A226541-100G A226541-10G A226541-25G A226541-500G A226541-5G A226541-1000G
77261-426EA 49.44 USD
77261-426 77261-428 77261-430 77261-432 77261-434 77746-330
N-(tert-Butoxycarbonyl)-4-aminobutyric acid 97%
N-(tert-Butoxycarbonyl)-4-aminobutyric acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
MDL Number: MFCD00037313
CAS Number: 57294-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR