H-Deg-OH 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-ethylbutanoic acid, H-Deg-OH, 3-Aminopentane-3-carboxylic acid

A226371-10G A226371-1G A226371-250MG A226371-5G
77275-628EA 240 USD
77275-628 77275-630 77275-632 77275-634
H-Deg-OH 97%
H-Deg-OH

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
MDL Number: MFCD01737173
CAS Number: 2566-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR