Danoprevir 99%

Supplier: Ambeed
Warning

Synonyms: ITMN-191, (2R,6S,13aS,14aR,16aS,Z)-6-((tert-butoxycarbonyl)amino)-14a-((cyclopropylsulphonyl)carbamoyl)-5,16-dioxo-1,2,3,5,6,7,8,9,10,11,13a,14,14a,15,16,16a-hexadecahydrocyclopropa[e]pyrrolo[1,2-a][1,4]diazacyclopentadecin-2-yl 4-fluoroisoindoline-2-carboxylate, RG7227

A226220-10MG A226220-1MG A226220-5MG
77237-746EA 77 USD
77237-746 77237-748 77237-750
Danoprevir 99%
Danoprevir

Precaución: for Research Use Only

Formula: C₃₅H₄₆FN₅O₉S
MDL Number: MFCD16038038
CAS Number: 850876-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR