Benzyl (2-sulfamoylethyl)carbamate 98%

Supplier: Ambeed
Warning

A225693-1G A225693-250MG A225693-5G
77261-408EA 8 USD
77261-408 77261-410 77261-412
Benzyl (2-sulfamoylethyl)carbamate 98%
Benzyl (2-sulfamoylethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₄S
MDL Number: MFCD28098092
CAS Number: 136027-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR