Saccharin 1-methylimidazole ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzo[d]isothiazol-3(2H)-one 1,1-dioxide compound with 1-methyl-1H-imidazole (1:1)
A225676-100MG
A225676-10MG
A225676-1MG
A225676-25MG
A225676-50MG
77261-398EA
53.56
USD
77261-398
77261-400
77261-402
77261-404
77261-406
Saccharin 1-methylimidazole ≥98%
Saccharin 1-methylimidazole
Precaución: for Research Use Only
Formula:
C₁₁H₁₁N₃O₃S MW: 265.29 g/mol |
MDL Number:
MFCD31657257 CAS Number: 482333-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...