3-Fluoro-4-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-4-methoxyphenyl)boronic acid, 3-Fluoro-4-methoxybenzeneboronic acid

A225640-100G A225640-10G A225640-1G A225640-25G A225640-5G
77275-612EA 134 USD
77275-612 77275-614 77275-616 77275-618 77275-620
3-Fluoro-4-methoxyphenylboronic acid 97%
3-Fluoro-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD00807404
CAS Number: 149507-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR