2-(3-Ethyl-4-oxothiazolidin-2-ylidene)malononitrile 98%

Supplier: Ambeed
Warning

A225550-100MG A225550-1G A225550-250MG
77261-392EA 15 USD
77261-392 77261-394 77261-396
2-(3-Ethyl-4-oxothiazolidin-2-ylidene)malononitrile 98%
2-(3-Ethyl-4-oxothiazolidin-2-ylidene)malononitrile

Precaución: for Research Use Only

Formula: C₈H₇N₃OS
MDL Number: MFCD30534266
CAS Number: 623558-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR