2-Phenyl-1,3,4-oxadiazole 95%

Supplier: Ambeed
Warning

A225522-1G A225522-250MG A225522-5G
77275-602EA 149 USD
77275-602 77275-604 77275-606
2-Phenyl-1,3,4-oxadiazole 95%
2-Phenyl-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD00491613
CAS Number: 825-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR