SC66 98%

Supplier: Ambeed
Warning

Synonyms: (2E,6E)-2,6-Bis(pyridin-4-ylmethylene)cyclohexanone

A225521-100MG A225521-10MG A225521-250MG A225521-50MG A225521-5MG
77237-718EA 195 USD
77237-718 77237-720 77237-722 77237-724 77237-726 77237-728 77237-730
SC66 98%
SC66

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O
MDL Number: MFCD05025493
CAS Number: 871361-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR