5-Bromo-6-fluoropyridin-3-ol 95%

Supplier: Ambeed
Warning

A225420-1G A225420-1MG A225420-250MG
77275-596EA 427.45 USD
77275-596 77275-598 77275-600
5-Bromo-6-fluoropyridin-3-ol 95%
5-Bromo-6-fluoropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃BrFNO
MW: 191.986 g/mol
MDL Number: MFCD08061303
CAS Number: 186593-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR