2,3-Dimethylbutanoic acid 95%

Supplier: Ambeed
Warning

A225321-100MG A225321-1G A225321-250MG
77249-474EA 54.59 USD
77249-474 77249-476 77249-478
2,3-Dimethylbutanoic acid 95%
2,3-Dimethylbutanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00045888
CAS Number: 14287-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR