D-(-)-Valine 97%

Supplier: Ambeed
Warning

A225204-100G A225204-10G A225204-25G A225204-500G A225204-1000G
77288-274EA 16 USD
77288-274 77288-276 77288-278 77288-280 77748-368
D-(-)-Valine 97%
D-(-)-Valine

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD00064219
CAS Number: 640-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR