2,3-Difluorophenetole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Ethoxy-2,3-difluorobenzene, 2,3-Difluoroethoxybenzene

A225181-100G A225181-10G A225181-1G A225181-25G A225181-500G A225181-5G
77249-462EA 28 USD
77249-462 77249-464 77249-466 77249-468 77249-470 77249-472
2,3-Difluorophenetole 98%
2,3-Difluorophenetole

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.15 g/mol
MDL Number: MFCD07368737
CAS Number: 121219-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR