5-Bromo-6-fluoropyridin-3-amine 97%

Supplier: Ambeed
Warning

A224994-1G A224994-250MG A224994-5G
77261-384EA 48 USD
77261-384 77261-386 77261-388
5-Bromo-6-fluoropyridin-3-amine 97%
5-Bromo-6-fluoropyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MDL Number: MFCD09835254
CAS Number: 209328-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR