4-Fluorobenzenesulfonic Acid 95%

Supplier: Ambeed
Warning

A224457-1G A224457-250MG A224457-5G
77288-268EA 56 USD
77288-268 77288-270 77288-272
4-Fluorobenzenesulfonic Acid 95%
4-Fluorobenzenesulfonic Acid

Precaución: for Research Use Only

Formula: C₆H₅FO₃S
MDL Number: MFCD06658253
CAS Number: 368-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR