3',5'-Bis(trifluoromethyl)propiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Bis(trifluoromethyl)phenyl)propan-1-one

A224352-100G A224352-10G A224352-1G A224352-25G A224352-5G
77275-576EA 283.25 USD
77275-576 77275-578 77275-580 77275-582 77275-584
3',5'-Bis(trifluoromethyl)propiophenone 98%
3',5'-Bis(trifluoromethyl)propiophenone

Precaución: for Research Use Only

Formula: C₁₁H₈F₆O
MW: 270.17 g/mol
MDL Number: MFCD00009911
CAS Number: 85068-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR