2-Hydroxy-3-methyl-5-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A224281-1G
A224281-250MG
A224281-5G
77261-372EA
40.17
USD
77261-372
77261-374
77261-376
2-Hydroxy-3-methyl-5-nitrobenzoic acid 97%
2-Hydroxy-3-methyl-5-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇NO₅ MW: 197.14 g/mol |
MDL Number:
MFCD13704881 CAS Number: 54674-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...