Potassium (Z)-3-cyano-3-(thiophen-2-yl)acrylate 97%

Supplier: Ambeed
Warning

A224277-100MG A224277-250MG
77237-708EA 15.45 USD
77237-708 77237-710
Potassium (Z)-3-cyano-3-(thiophen-2-yl)acrylate 97%
Potassium (Z)-3-cyano-3-(thiophen-2-yl)acrylate

Precaución: for Research Use Only

Formula: C₈H₄KNO₂S
MW: 217.29 g/mol
MDL Number: MFCD29924754
CAS Number: 912368-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR