4-Amino-1-((2R,4S,5R)-5-(((tert-butyldimethylsilyl)oxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one 97%

Supplier: Ambeed
Warning

A224253-100MG A224253-250MG
77249-450EA 26.78 USD
77249-450 77249-452
4-Amino-1-((2R,4S,5R)-5-(((tert-butyldimethylsilyl)oxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one 97%
4-Amino-1-((2R,4S,5R)-5-(((tert-butyldimethylsilyl)oxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₅H₂₆IN₃O₄Si
MW: 467.37 g/mol
MDL Number: MFCD09264239
CAS Number: 666848-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR