Benzo[b]naphtho[1,2-d]thiophen-5-ylboronic acid 97%

Supplier: Ambeed
Warning

A224183-250MG A224183-1G
77249-448EA 11.33 USD
77249-448 77615-864
Benzo[b]naphtho[1,2-d]thiophen-5-ylboronic acid 97%
Benzo[b]naphtho[1,2-d]thiophen-5-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₁BO₂S
MW: 278.13 g/mol
MDL Number: MFCD30502871
CAS Number: 1447709-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR