4-Methyl-3-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methoxy-4-methyl-3-nitrobenzene, 4-Methoxy-2-nitrotoluene, Methyl 3-nitro-p-tolyl ether

A224020-10G A224020-1G A224020-25G A224020-5G
77249-440EA 23 USD
77249-440 77249-442 77249-444 77249-446
4-Methyl-3-nitroanisole 98%
4-Methyl-3-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD00007173
CAS Number: 17484-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR