2-Chloro-5-(trifluoromethyl)benzimidazole 95%

Supplier: Ambeed
Warning

A223908-100MG A223908-1G A223908-250MG
77261-362EA 20 USD
77261-362 77261-364 77261-366
2-Chloro-5-(trifluoromethyl)benzimidazole 95%
2-Chloro-5-(trifluoromethyl)benzimidazole

Precaución: for Research Use Only

Formula: C₈H₄ClF₃N₂
MDL Number: MFCD12963759
CAS Number: 86604-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR