2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-N-(4-(trifluoromethyl)phenyl)acetamide

A223623-1G A223623-250MG A223623-5G
77261-356EA 53.56 USD
77261-356 77261-358 77261-360
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide 98%
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂O
MW: 228.17 g/mol
MDL Number: MFCD05669245
CAS Number: 24522-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR