2-(Aminomethyl)-4-chloroaniline 97%

Supplier: Ambeed
Danger

A223524-1G A223524-250MG
77237-684EA 96.82 USD
77237-684 77237-686
2-(Aminomethyl)-4-chloroaniline 97%
2-(Aminomethyl)-4-chloroaniline

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.61 g/mol
MDL Number: MFCD06212615
CAS Number: 108047-39-8
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR