3-Fluoro-4-methylbenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)-2-fluoro-1-methylbenzene

A223458-10G A223458-1G A223458-5G
77275-548EA 83 USD
77275-548 77275-550 77275-552
3-Fluoro-4-methylbenzyl bromide 97%
3-Fluoro-4-methylbenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.05
MDL Number: MFCD01631534
CAS Number: 145075-44-1
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR