(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-[2-Nitro-4-(trifluoromethyl)phenyl]acetic acid

A223410-10G A223410-1G A223410-25G A223410-5G
77261-348EA 50 USD
77261-348 77261-350 77261-352 77261-354
(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid 98%
(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆F₃NO₄
MDL Number: MFCD00091534
CAS Number: 1735-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR