Imidazole-4,5-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A223300-100G A223300-10G A223300-25G A223300-500G A223300-5G
77275-540EA 51.01 USD
77275-540 77275-542 77275-544 77275-546 77748-366
Imidazole-4,5-dicarboxylic acid 95%
Imidazole-4,5-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄
MDL Number: MFCD00005200
CAS Number: 570-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR