2-Chloro-5-nitro-1H-1,3-benzimidazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-nitro-1H-benzo[d]imidazole

A223144-1G A223144-250MG
77261-344EA 135.96 USD
77261-344 77261-346
2-Chloro-5-nitro-1H-1,3-benzimidazole 95%
2-Chloro-5-nitro-1H-1,3-benzimidazole

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MW: 197.58 g/mol
MDL Number: MFCD00769824
CAS Number: 5955-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR