Ethyl-2-methylthiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

A223135-10G A223135-1G A223135-250MG A223135-25G A223135-5G
77275-524EA 286 USD
77275-524 77275-526 77275-528 77275-530 77275-532
Ethyl-2-methylthiazole-5-carboxylate 98%
Ethyl 2-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MDL Number: MFCD03840442
CAS Number: 79836-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR