2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-dicarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A223108-1G
A223108-250MG
77237-680EA
130
USD
77237-680
77237-682
2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-dicarboxylic acid 98%
2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₆H₈F₆O₄ |
MDL Number:
MFCD30533922 CAS Number: 89803-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...