1-Chloro-3,5-dimethoxybenzene 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-1,3-dimethoxybenzene

A223029-100G A223029-10G A223029-1G A223029-25G A223029-5G
77237-670EA 339.9 USD
77237-670 77237-672 77237-674 77237-676 77237-678
1-Chloro-3,5-dimethoxybenzene 97%
1-Chloro-3,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.61 g/mol
MDL Number: MFCD00008382
CAS Number: 7051-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR