2',5'-Difluoropropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Difluorophenyl)propan-1-one

A222800-1G A222800-25G A222800-5G
77249-420EA 10.3 USD
77249-420 77249-422 77249-424
2',5'-Difluoropropiophenone 98%
2',5'-Difluoropropiophenone

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.16 g/mol
MDL Number: MFCD00015509
CAS Number: 29112-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR