2-Chlorobenzothiazole-6-carbonitrile 95%

Supplier: Ambeed
Warning

A222793-250MG A222793-1G
77275-516EA 89.61 USD
77275-516 77748-362
2-Chlorobenzothiazole-6-carbonitrile 95%
2-Chlorobenzothiazole-6-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₃ClN₂S
MW: 194.6408 g/mol
MDL Number: MFCD09743933
CAS Number: 80945-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR