1-Chloro-4-iodobutane 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A222627-100G A222627-10G A222627-1G A222627-25G A222627-500G A222627-5G
77237-658EA 146.26 USD
77237-658 77237-660 77237-662 77237-664 77237-666 77237-668
1-Chloro-4-iodobutane 98% stabilized
1-Chloro-4-iodobutane

Precaución: for Research Use Only

Formula: C₄H₈ClI
MW: 218.46 g/mol
MDL Number: MFCD00039415
CAS Number: 10297-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR