2-Bromo-5-chloro-3-hydroxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-chloropyridin-3-ol

A222598-100MG A222598-1G A222598-250MG A222598-5G
77261-320EA 16.48 USD
77261-320 77261-322 77261-324 77593-440
2-Bromo-5-chloro-3-hydroxypyridine 95%
2-Bromo-5-chloro-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClNO
MW: 208.44 g/mol
MDL Number: MFCD09835245
CAS Number: 127561-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR